Face Shields
Oberon Gold Reflective Faceshield for high radiant heat application.
The Heat Reflective Series of faceshields is manufactured using space-aged technology. The face shields are coated with a mirror-like surface of 24K Gold to reflect the radiant energy away from the user’s face.
Headgear is ratchet adjustable and comfortable. Ideal for shop, industrial and laboratory use.