Gold Heat Reflective Face Shields, Oberon
Supplier: Sellstrom
125R 025
89370-940EA 178.2 USD
89370-940 80700-054 89370-941
Gold Heat Reflective Face Shields, Oberon
Face Shields
The Oberon Gold Reflective Faceshield is designed for high radiant heat applications.
  • Heat reflective series of faceshields is manufactured using space-age technology
  • Coated with a mirror-like surface of 24K Gold to reflect the radiant energy away from the user’s face
  • Headgear is ratchet adjustable and comfortable
  • Ideal for shop, industrial, and laboratory use
Ordering information: Contact your Avantor sales representative for more information.