HPI-4 (Ciliobrevin A), Powder

Supplier: SELLECK CHEMICALS LLC MS
Warning

Synonyms: Hedgehog Pathway Inhibitor 4, Ciliobrevin A, 2,4-Dichloro-a-(3,4-dihydro-4-oxo-2(1H)-quinazolinylidene)-β-oxo-benzenepropanenitrile, 3-(2,4-Dichlorophenyl)-3-oxo-2-(4-oxo-3,4-dihydroquinazolin-2(1H)-ylidene)propanenitrile

S8249-10MG S8249-50MG
103548-726EA 126.15 USD
103548-726 103548-728 103548-730
HPI-4 (Ciliobrevin A), Powder
HPI-4 (Ciliobrevin A)

HPI-4 (Ciliobrevin A) is a hedgehog (Hh) pathway antagonist. It blocks Sonic hedgehog (Shh)-induced Hh pathway activation (IC50 = 7 μM) downstream of Smo.

Formula: C₁₇H₉Cl₂N₃O₂
MW: 358.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01911757
CAS Number: 302803-72-1
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR