CB1954 (Tretazicar)

Supplier: SELLECK CHEMICALS LLC MS

Synonyms: 5-(Aziridin-1-yl)-2,4-dinitrobenzamide, Tretazicar, 5-Aziridinyl-2,4-dinitrobenzamide

S7829-10MG S7829-50MG S7829-200MG
103547-212EA 181.54 USD
103547-212 103547-214 103547-216
CB1954 (Tretazicar)
CB1954 (Tretazicar)

CB1954(Tretazicar) is a anticancer prodrug that is converted in the presence of the enzyme NQO2 and co-substrate caricotamide ( EP-0152R) (EP) into a potent cytotoxic bifunctional alkylating agent. It can be activated by NAD(P)H quinone oxidoreductase 2.

Formula: C₉H₈N₄O₅
Storage Temperature: Freezer
MDL Number: MFCD00869490
CAS Number: 21919-05-1
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR