JSH-23 ≥98%

Supplier: Cayman Chemical Company

Synonyms: 4-Methyl-N1-(3-phenylpropyl)-1,2-phenylenediamine, JSH-23, 4-Methyl-N1-(3-phenylpropyl)benzene-1,2-diamine

15036-1 15036-5 15036-10 15036-25
76293-006EA 63.92 USD
76293-006 76293-008 76293-010 76293-012
JSH-23 ≥98%
JSH-23

  • An inhibitor of NF-κB translocation


An inhibitor of NF-κB, blocking its translocation into the nucleus (IC50 = 7.1 µM); does not affect degradation of IκB; interferes with NF-κB-mediated gene expression and apoptosis in RAW 264.7 cells treated with LPS

Formula: C₁₆H₂₀N₂
MW: 240.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09753595
CAS Number: 749886-87-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae8fffebff4d4f095d53a6db5045b56971bb98d9&vid=ae8fffebff4d4f095d53a6db5045b56971bb98d9&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d4425efb15c8ec572238ebbbc419c018ee5be72&vid=2d4425efb15c8ec572238ebbbc419c018ee5be72&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=996c7bcb6cf8a40dcd360dde5dc6a1f9a28c0c61&vid=996c7bcb6cf8a40dcd360dde5dc6a1f9a28c0c61&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥98%
Stability ≥ 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR