6-Carboxyfluorescein
Supplier: Anaspec Inc
Synonyms:
3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid, 6-FAM
AS-81004
AS-81004-1
103010-760EA
125.69
USD
103010-760
103010-762
6-Carboxyfluorescein
6-Carboxyfluorescein
6-FAM is another isomer of carboxyfluorescein. It is mainly used in sequencing of nucleic acids and labeling nucleotides.
- Spectral properties: Abs/Em = 495/517 nm
- Solvent system: DMF or DMSO
- Appearance: Red solid
- Storage Temperature: -20°C desiccated and protected from light
- Shipping condition: RT
Formula:
C₂₁H₁₂O₇ MW: 376.32 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00036873 CAS Number: 3301-79-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...