Anti-IgE Rat Antibody (RPE (R-Phycoerythrin)) [clone: 23G3]

Supplier: Southern Biotechnology
1130-09L 1130-09
100242-792EA 438.08 USD
100242-792 100242-790
Anti-IgE Rat Antibody (RPE (R-Phycoerythrin)) [clone: 23G3]
Antibodies
PE Anti-Mouse IgE antibody for use in flow cytometry assays.

Type: Secondary
Antigen: IgE
Clonality: Monoclonal
Clone: 23G3
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR