Human Recombinant FasL (from HEK293 cells)
Supplier: ADIPOGEN CORP MS
AG-40B-0130-C010 AG-40B-0130-3010
102980-234EA 443.08 USD
102980-234 102980-236
Human Recombinant FasL (from HEK293 cells)
Proteins and Peptides
MultimericFasL™ is a high activity construct in which two trimeric FasL are artificially linked via the collagen domain of ACRP30. This construct very effectively mimics the natural membrane-assisted aggregation of FasL in vivo. FasL is a cytokine that binds to TNFRSF6/Fas, a receptor that transduces the apoptotic signal into cells. It is involved in cytotoxic T cell mediated apoptosis and in T cell development.