Human Recombinant Irisin (from HEK293 Cells)
Supplier: ADIPOGEN CORP MS
AG-40B-0115-C010 AG-40B-0115-5010
102980-182EA 180 USD
102980-182 102980-184
Human Recombinant Irisin (from HEK293 Cells)
Proteins and Peptides
Irisin is a recently described exercise-induced hormone secreted by skeletal muscle in mice and humans. Irisin activates beige fat cells (beige cells have a gene expression pattern distinct from either white or brown fat and are preferentially sensitive to the polypeptide hormone Irisin). Irisin is cleaved from the type I membrane protein FNDC5 and improves systemic metabolism by increasing energy expenditure.