Chaetoviridin A (from Chaetomium sp.) ≥96% (by HPLC)
Supplier: ADIPOGEN CORP MS
BVT-0419-M001
BVT-0419-M005
102988-978EA
159.23
USD
102988-978
102988-980
Chaetoviridin A (from Chaetomium sp.) ≥96% (by HPLC)
Chaetoviridin A
Chaetoviridin A is an azaphilone antibiotic.
- Anti-fungal compound
- Improves blood lipid levels
- Monoamine oxidase inhibitor
Anti-fungal compound (phytopathogenic fungi). Cholesteryl ester transfer protein (CETP) inhibitor. Improves blood lipid levels.
Yellow to orange powder
Soluble in DMSO, methanol, chloroform or acetone
Storage Temperature:
Freezer |
CAS Number:
128252-98-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...