Hormaomycin (from Streptomyces griseoflavus) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0107-C500 BVT-0107-M001
102988-888EA 346.15 USD
102988-888 102988-890
Hormaomycin (from Streptomyces griseoflavus) ≥98% (by HPLC)
Hormaomycin

Hormaomycin is a potent narrow-spectrum antibiotic against coryneform actinomycetes (MIC value 88 pM).


  • Antibiotic
  • Shows hormonal activity
  • Rare type of natural product


Antibacterial agent (selective). Quorum sensing modulator. Inducer of morphological differentiation in actinomycetes. Stimulator of antibiotic production. Inducer of aerial mycelium formation in various actinomycetes. Antimalarial (in vitro). Rare type of natural product, regulating bacterial behavior and independently function in a defensive antibiotic role.

Formula: C₅₅H₆₉N₁₀O₁₄Cl
CAS Number: 121548-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR