Quinolactacin A1 : Quinolactacin A2
Supplier: ADIPOGEN CORP MS
AG-CN2-0164-M001 AG-CN2-0164-M005
102987-434EA 304.62 USD
102987-434 102987-436
Quinolactacin A1 : Quinolactacin A2
Quinolactacin A1 : Quinolactacin A2
Antibiotic
  • Quinolactacin A1: 386211-68-3
  • Quinolactacin A2: 386211-69-4
Acetylcholinesterase (AChE) inhibitor. Anti-inflammatory compound. Tumor necrosis factor (TNF) inhibitor.
CAS Number: 386211-68-3 (Quinolactacin A1) - 386211-69-4 (Quinolactacin A2)