GSK25 ≥95%
Supplier: ADIPOGEN CORP MS
SYN-1124-M001
SYN-1124-M005
SYN-1124-M010
102992-206EA
283.85
USD
102992-206
102992-208
102992-210
GSK25 ≥95%
GSK25
GSK-25 maintains good selectivity against a panel of 31 kinases, as well as RSK1 and p70S6K (RSK1 IC(50) of 398nM, p70S6K IC(50) of 1000nM), and a dramatically improved P450 profile (
Formula:
C₂₄H₁₆Cl₂F₂N₆O Storage Temperature: Freezer |
MDL Number:
MFCD18782716 CAS Number: 874119-56-9 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...