S-99 ≥95%

Supplier: ADIPOGEN CORP MS
Warning

SYN-1119-M001 SYN-1119-M005 SYN-1119-M010
102992-168EA 283.85 USD
102992-168 102992-170 102992-172
S-99 ≥95%
S-99

Selective ASKI inhibitor. S-99 inhibits LPS-induced TNFalpha release assay in vivo.

Formula: C₁₆H₁₅F₃N₆O
Storage Temperature: Freezer
CAS Number: 1124381-69-6

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR