YM201636 hydrochloride ≥95%

Supplier: ADIPOGEN CORP MS

SYN-1094-M001 SYN-1094-M005 SYN-1094-M010 SYN-1094-M050
102991-996EA 149.54 USD
102991-996 102991-998 102992-000 102992-002
YM201636 hydrochloride ≥95%
YM201636 hydrochloride

YM-201636 is a cell-permeable and selective inhibitor of PIKfyve (IC(50) of 33nM). It disrupts retroviral budding from cells, apparently by interfering with the endosomal sorting complex required for transport (ESCRT) machinery. In adipocytes, YM-201636 also inhibits basal and insulin-activated 2-deoxyglucose uptake (IC(50) of 54nM).

Formula: C₂₅H₂₁N₇O₃·HCl
Storage Temperature: Freezer
CAS Number: 371942-69-7

Order Now

Specification Test Results

Purity >95%
Appearance/Color Solid.
Solubility Soluble in DMSO. Slightly soluble (<1mg/ml) in ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR