Amuvatinib ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: N-(Benzo[d][1,3]dioxol-5-ylmethyl)-4-(benzofuro[3,2-d]pyrimidin-4-yl)piperazine-1-carbothioamide, MP-470

SYN-1056-M001 SYN-1056-M005 SYN-1056-M010 SYN-1056-M050
102991-684EA 149.54 USD
102991-684 102991-686 102991-688 102991-690
Amuvatinib ≥95%
Amuvatinib

Amuvatinib is a potent, novel and orally active multi-targeted tyrosine kinase inhibitor, which inhibits a number of validated tumor targets including c-kit, PDGFRalpha and flt3, with IC(50) values of 10nM, 40nM and 81nM respectively.

Formula: C₂₃H₂₁N₅O₃S
Storage Temperature: Freezer
MDL Number: MFCD16038298
CAS Number: 850879-09-3

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR