(±)-Sodium-2,3-dihydroxyisovalerate hydrate ≥96%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
Sodium 2,3-dihydroxy-3-methylbutanoate hydrate
CDX-S0055-M020
CDX-S0055-M100
102991-014EA
123.23
USD
102991-014
102991-016
(±)-Sodium-2,3-dihydroxyisovalerate hydrate ≥96%
(±)-Sodium-2,3-dihydroxyisovalerate hydrate
Racemic form of intermediate in L-valine catabolism and anabolism.
Formula:
C₅H₉NaO₄·H₂O Storage Temperature: Freezer |
MDL Number:
MFCD12912381 CAS Number: 74294-97-6 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White to off-white powder. |
Solubility | Soluble in water. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...