Ferimzone ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Meferimzone

CDX-F0081-M100 CDX-F0081-M500
102989-998EA 210.46 USD
102989-998 102990-000
Ferimzone ≥98%
Ferimzone

Systemic fungicide developed for the control of rice diseases that exhibits strong antifungal activity against many plant-pathogenic fungi. Antifungal activity does not affect the synthesis of RNA, DNA, protein, cell walls, or lipid components. Inhibits mycelial growth, but does not inhibit spore germination until a germ tube had formed. No effect of ferimzone was observed on the cell wall architecture of mycelia. Ferimzone causes leakage of some electrolytes from mycelia, which suggests that it affects or disrupts membrane function. Compound can be used as analytical reference material.

Formula: C₁₅H₁₈N₄
Storage Temperature: Freezer
CAS Number: 89269-64-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Colorless or white powder/crystals.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR