Flumetsulam ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: DE-498

CDX-F0080-M100 CDX-F0080-G001
102989-994EA 110.77 USD
102989-994 102989-996
Flumetsulam ≥98%
Flumetsulam

Highly active herbicide of the triazolopyrimidine sulfonanilide class. Flumetsulam acts by inhibiting acetolactate synthase. Flumetsulam exhibits broad spectrum activity on broadleaf weed species with selectivity to several major agronomic crops in soil and foliar applications. Compound can be used as analytical reference material.

Formula: C₁₂H₉F₂N₅O₂S
Storage Temperature: Freezer
MDL Number: MFCD04089841
CAS Number: 98967-40-9
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color White to light Beige powder/crystals.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR