ASPT ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: trans-​4-​[4-​(N-​Ethyl-​2-​hydroxyethylamino)​styryl]​-​1-​methylpyridinium tetraphenylborate

CDX-E0006-M100 CDX-E0006-G001
102989-876EA 203.54 USD
102989-876 102989-878
ASPT ≥95%
ASPT

(Aminostyryl)pyridinium hemicyanine dye with solvent-dependent one- and two-photon emission1.

Formula: C₄₂H₄₃BN₂O
Storage Temperature: Freezer
CAS Number: 159721-38-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPCE)
Appearance/Color Solid.
Solubility Soluble in acetonitrile.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR