DAR-4M ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Diaminorhodamine-4M

CDX-D0206-M001 CDX-D0206-M005
102989-668EA 351.69 USD
102989-668 102989-670
DAR-4M ≥98%
DAR-4M

Photo-stable fluorescent probe (Ex: ~560nm; Em: 575nm) for the detection of nitric oxide (NO) in presence of oxygen, resulting in a triazolo-rhodamine analog (DAR-4M T) that exhibits about 840-fold greater fluorescence quantum efficiency. Suitable probe for monitoring RNS production, but not NO alone. Stable in a pH range of 4-12. Detection limit ~10nM.

Formula: C₂₅H₂₆N₄O₃
Storage Temperature: Freezer
CAS Number: 339527-79-6

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (HPLC)
Appearance/Color Reddish-violet powder.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR