DAR-M ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Diaminorhodamine-M

CDX-D0121-M001 CDX-D0121-M005
102987-816EA 351.69 USD
102987-816 102987-818
DAR-M ≥98%
DAR-M

Photo-stable fluorescent probe (Ex: ~565nm; Em: 580nm) for the detection of nitric oxide (NO).

Formula: C₂₉H₃₄N₄O₃
Storage Temperature: Freezer
CAS Number: 261351-44-4

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (NMR)
Appearance/Color Violet powder.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR