7-(Diethylamino)-3-(imidazol-1-ylcarbonyl)-1-benzopyran-2-one ≥97%

Supplier: ADIPOGEN CORP MS

Synonyms: 7-Diethylaminocoumarin-3 carboxylic acid imidazolide

CDX-D0071-M100 CDX-D0071-G001
102987-746EA 319.85 USD
102987-746 102987-748
7-(Diethylamino)-3-(imidazol-1-ylcarbonyl)-1-benzopyran-2-one ≥97%
7-(Diethylamino)-3-(imidazol-1-ylcarbonyl)-1-benzopyran-2-one

fluorescent reagent for use in circular dichroic exciton method for derivatisation of hydroxyl-groups under mild conditions. The absorption at 406 nm is futher in the red than for reagents like naphthoylimidazole. The reagent offers high sensitivity through a high molar extinction coefficient.

Formula: C₁₇H₁₇N₃O₃
Storage Temperature: Freezer
CAS Number: 261943-47-9

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR