PyTC2 ≥85%
Supplier: ADIPOGEN CORP MS
|
Danger
|
Synonyms:
1-(Dichloro-1,3,5-triazinyl)-pyrene
Fluorescent probe with high quantum efficiency, exhibiting single exponential decay kinetics. The radiative fluorescence lifetime is almost constant and independent on molecular surroundings allowing this probe to be utilized in a range of microsurroundings with exciting radiation at a constant wavelength (in the blue spectral range). Its fluorescence emission demonstrates a large solvatochromic shift which changes with solvent polarity. For example, the maximum shifts from 435 nm in low polarity n-heptane to 492 nm in high polarity acetonitrile. This product is only sparingly soluble in water
Formula:
C₁₉H₉Cl₂N₃ Storage Temperature: Refrigerator |
CAS Number:
3224-36-0 UN: 2811 ADR: 6.1,III |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White powder. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...