DACB-CN ≥90%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 4-(7-Diethylaminocoumarin-3-yl)benzoyl cyanide

CDX-D0016-M010 CDX-D0016-M100
102989-618EA 257.54 USD
102989-618 102989-620
DACB-CN ≥90%
DACB-CN

Analytical reagent for chromatography. Derivatization reagents.

Formula: C₂₁H₁₈N₂O₃
Storage Temperature: Freezer
CAS Number: 203256-20-6

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 90% (HPCE)
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR