5-Carboxytetramethylrhodamine (5-TAMRA) ≥97%

Supplier: ADIPOGEN CORP MS

Synonyms: 5-TAMRA, 5-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-5-carboxybenzoate, 2-[3,6-Bis(dimethylamino)xanthen-10-ium-9-yl]-5-carboxybenzoate, TAMRA, 5-isomer

CDX-C0055-M025 CDX-C0055-M250
102987-638EA 210.46 USD
102987-638 102987-640
5-Carboxytetramethylrhodamine (5-TAMRA) ≥97%
5-Carboxytetramethylrhodamine (5-TAMRA)

Free acid form of 5-carboxytetramethylrhodamine single isomer. Widely used fluorophore for preparing bioconjugates, especially fluorescent antibody and avidin derivatives used in immunochemistry. Used as a dye for oligonucleotide labeling and automated DNA sequencing applications.

Formula: C₂₅H₂₂N₂O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00269769
CAS Number: 91809-66-4

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (HPLC)
Appearance/Color Solid.
Solubility Soluble in methanol, DMSO or DMF.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR