AMCA-X-N-succinimidyl ester ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: AMCA-X, SE, 6-((7-Amino-4-methylcoumarin-3-acetyl)amino) hexanoic acid, N-succinimidyl ester

CDX-A0074-M010 CDX-A0074-M100
102989-184EA 318.46 USD
102989-184 102989-186
AMCA-X-N-succinimidyl ester ≥95%
AMCA-X-N-succinimidyl ester

AMCA-X SE is an amine-reactive, UV-excitable, blue fluorescent dye.

Formula: C₂₂H₂₅N₃O₇
MW: 443.45 g/mol
Storage Temperature: Freezer
CAS Number: 216309-02-3

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Light yellow powder.
Solubility Soluble in DMSO or DMF.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR