Sterigmatocystin ≥98%

Supplier: ADIPOGEN CORP MS
Danger

BVT-0171-M001 BVT-0171-M005
102988-902EA 69.23 USD
102988-902 102988-904
Sterigmatocystin ≥98%
Sterigmatocystin

Sterigmatocystin is an intermediate of the biosynthetic pathway to Aflatoxin B1.


  • DNA synthesis inhibitor
  • Cytotoxic, carcinogenic and mutagenic
  • Mycotoxin


Anti-cancer compound. Acyl-CoA:cholesterol acyltransferase 2 (ACAT2) inhibitor. Shown to induce apoptosis in human peripheral lymphocytes and necrosis in rat liver. Induces sister chromatid exchanges in murine bone marrow cells.


Pale Yellow powder


Soluble in DMSO, ethanol, methanol, pyridine or acetone

Formula: C₁₈H₁₂O₆
Storage Temperature: Freezer
CAS Number: 10048-13-2
UN: 3462
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Isolated from Aspergillus sp. (strain WDMH51).
Purity 98% (HPLC; NMR)
Appearance/Color Pale yellow powder.
Solubility Soluble in DMSO, ethanol, methanol, pyridine or acetone.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR