(S)-Roscovitine ≥99%

Supplier: ADIPOGEN CORP MS

Synonyms: 6-Benzylamino-2-(S)-[(1-ethyl)-2-hydroxyethylamino]-9-isopropylpurine

AG-MR-C0002-M005 AG-MR-C0002-M025
102987-462EA 138.46 USD
102987-462 102987-464
(S)-Roscovitine ≥99%
(S)-Roscovitine

(S)-enantiomer of (R)-roscovitine . Potent selective cyclin-dependent kinase 1 (CDK1) inhibitor. Potential neuroprotectant for stroke. Crosses the blood brain barrier. Inhibits CDK5, consequently blocking hypoxia-induced apoptosis in neurons. Potential anti-inflammatory compound. Potential antidiabetic compound.

Formula: C₁₉H₂₆N₆O
Storage Temperature: Freezer
CAS Number: 186692-45-5

Order Now

Specification Test Results

Purity 99% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO, ethanol or methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR