2,5-Dimethyl-celecoxib ≥99%
Supplier: ADIPOGEN CORP MS
|
Danger
|
Synonyms:
DMC
Close structural analog of the selective cyclooxygenase-2 (COX-2) inhibitor celecoxib, that lacks COX-2 inhibitory function. Anti-proliferative, anti-tumorigenic and anti-angiogenic. Potent apoptosis inducer in many cancer cell lines. Down-regulates the expression of survivin, cyclins, MMPs and inhibits cyclin-dependent kinase activity. Modulates PDK-1, AKT, GSK3beta, p70 S6K, PKA and MAPKAP-K1alpha. Reduces phosphorylation of ERK1/2 but not AKT T-308 or AKT S-473. Increases intracellular free calcium levels and potently triggers the endoplasmatic reticulum stress response (ESR), activating ER stress-associated proteins GRP78/BiP, CHOP/GADD153 and caspase-4.
Formula:
C₁₈H₁₆F₃N₃O₂S Storage Temperature: Freezer |
MDL Number:
MFCD19443858 CAS Number: 457639-26-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity |
|
Appearance/Color | White solid. |
Solubility | Soluble in DMSO, methanol or dichloromethane. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...