Actinonin ≥97%
Supplier: ADIPOGEN CORP MS
AG-CN2-0161-M005
AG-CN2-0161-M025
102987-422EA
135.69
USD
102987-422
102987-424
Actinonin ≥97%
Actinonin
Antibiotic
Source/Host: Synthetic.
Purity: >97.0% (HPLC)
Appearance/Color: White solid.
Solubility: Soluble in DMSO or ethanol.
Aminopeptidase M, N and leucine and alanine aminopeptidase inhibitor. Potent peptide deformylase (PDF) inhibitor. Inhibits MMP-1, MMP-2, MMP-3, MMP-7, MMP-8, MMP-9, MMP-10, MMP-12 and MMP-13, but not MMP-14. Anticancer compound. Antiproliferative. Shows antitumor activity in vitro and in vivo Apoptosis inducer. Meprin A inhibitor.
Storage Temperature:
Freezer |
CAS Number:
13434-13-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...