OM173-αA ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
AG-CN2-0158-M001
AG-CN2-0158-M005
102988-114EA
263.08
USD
102988-114
102988-116
OM173-αA ≥95%
OM173-alphaA
Antibiotic
Source/Host: Isolated from Streptomyces sp.
Purity: >95% (HPLC)
Appearance/Color: Yellow solid.
Solubility: Soluble in DMSO, ethanol or methanol.
Antimycoplasma. Antifungal compound. Anti-malarial agent. Ras-competitive non-CAAX mimetic type farnesyltransferase (FTase) inhibitor. Potential anticancer compound. Inducer of antiproliferative effects in tumor cell lines. Selective DNA (cytosine-5)-methyltransferase 3B (DNMT3B) inhibitor.
Storage Temperature:
Freezer |
CAS Number:
58286-56-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...