Pyridoxatin ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
AG-CN2-0123-C250
AG-CN2-0123-M001
102987-986EA
76.15
USD
102987-986
102987-988
Pyridoxatin ≥95%
Pyridoxatin
Antibiotic
Source/Host: Isolated from Albophoma yamanashiensis.
Purity: >95% (HPLC)
Appearance/Color: White solid.
Solubility: Soluble in DMSO or methanol.
Free radical scavenger. Lipid peroxidation inhibitor. Antimicrobial activity against Candida albicans. Gelatinase A (MMP-2) inhibitor. Anticancer. Cytotoxic. DNA synthesis inhibitor.
Storage Temperature:
Freezer |
CAS Number:
135529-30-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...