Penicillide ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Vermixocin A

AG-CN2-0122-C250 AG-CN2-0122-M001
102987-982EA 69.23 USD
102987-982 102987-984
Penicillide ≥95%
Penicillide

Plant growth inhibitor. Acyl-CoA-cholesterol acyltransferase (ACAT) inhibitor. Anticancer compound. Cytotoxic. Non-peptidic oxytocin receptor antagonist. Calpain inhibitor.

Formula: C₂₁H₂₄O₆
Storage Temperature: Freezer
CAS Number: 55303-92-9

Order Now

Specification Test Results

Source/Host Isolated from Penicillium sp.
Purity 95% (HPLC)
Appearance/Color Off-white solid.
Solubility Soluble in DMSO, ethanol or methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR