Harzianum A ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
AG-CN2-0117-C250
AG-CN2-0117-M001
102987-410EA
69.23
USD
102987-410
102987-412
Harzianum A ≥95%
Harzianum A
Antifungal. Anticancer compound. Cytotoxic to human cancer cell lines.
Formula:
C₂₃H₂₈O₆ Storage Temperature: Freezer |
CAS Number:
156250-74-7 |
Specification Test Results
Source/Host | Isolated from Trichoderma sp. |
Purity |
|
Appearance/Color | Off-white solid. |
Solubility | Soluble in DMSO, ethanol or methanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...