Aranorosin ≥95%

Supplier: ADIPOGEN CORP MS
Warning

AG-CN2-0114-C250 AG-CN2-0114-M001
102987-398EA 69.23 USD
102987-398 102987-400
Aranorosin ≥95%
Aranorosin

Antibiotic


Source/Host: Isolated from Gymnascella sp.
Purity: >95% (HPLC)
Appearance/Color: White solid.
Solubility: Soluble in DMSO, ethanol or methanol.


Antibacterial. Antifungal. Anticancer compound. Inhibits anti-apoptotic functions regulated by Bcl-2. Circumvents arbekacin (ABK)-resistance in MRSA by inhibiting the bifunctional enzyme AAC(6')/APH(2'').

Storage Temperature: Freezer
CAS Number: 117184-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR