N-Hydroxy-N'-phenyloctanediamide
Supplier: ADIPOGEN CORP MS
Synonyms:
N1-Hydroxy-N8-phenyloctanediamide, Suberoylanilide Hydroxamic Acid, Vorinostat
Vorinostat (SAHA, MK0683, Zolinza) inhibits the enzymatic activity of histone deacetylases HDAC1, HDAC2 and HDAC3 (Class I) and HDAC6 (Class II) at nanomolar concentrations (IC50
Formula:
C₁₄H₂₀N₂O₃ MW: 264.33 g/mol |
MDL Number:
MFCD00945317 CAS Number: 149647-78-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...