Xylylazo violet I

Supplier: ADIPOGEN CORP MS

Synonyms: 2-[2-Hydroxy-3-(2,4-xylylcarbamoyl)-1-naphthylazo]-1-phenol-4-sulphonic acid sodium salt, 1-Azo-2-hydroxy-3-(2,4-dimethylcarboxyanilido)naphthalene-1'-(2-hydroxybenzene-5-sulphonic acid) sodium salt, Xylidyl Blue I, 2-[2-Hydroxy-3-(2,4-xylylcarbamoyl)-1-naphthylazo]-1-phenol-4-sulfonic acid sodium salt, 1-Azo-2-hydroxy-3-(2,4-dimethylcarboxyanilido)naphthalene-1'-(2-hydroxybenzene-5-sulfonic acid) sodium salt

CDX-X0009-G001 CDX-X0009-G010 CDX-X0009-G025
102991-214EA 78.92 USD
102991-214 102991-216 102991-218
Xylylazo violet I
Xylylazo Violet I

Colorimetric reagent for Mg detection. The aqueous solution of XB-1 is red, and turns reddish-violet in the presence of Mg at pH 9 (maximum wavelength: 510 nm, molar absorptivity: 49,000, detection range 0.02-0.4 ppm).

Formula: C₂₅H₂₁N₃O₆S
MDL Number: MFCD00003890
CAS Number: 14936-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR