2,2':6',2''-Terpyridine
Supplier: ADIPOGEN CORP MS
Synonyms:
TERPY
CDX-T0113-G001
CDX-T0113-G005
102991-124EA
139.85
USD
102991-124
102991-126
2,2':6',2''-Terpyridine
2,2':6',2''-Terpyridine
Reagent for the spectrophotometric det. of Ag and Ru. Tridentate ligand that binds metals at three meridional sites giving two adjacent 5-membered MN2C2 chelate rings. It forms complexes with most transition metal ions. It is also a potent inducer of nitrate and nitric oxide (NO) synthase (iNOS) protein and mRNA production in the cultured murine macrophage RAW 264.7 cell line.
Formula:
C₁₅H₁₁N₃ MW: 233.27 g/mol |
MDL Number:
MFCD00006213 CAS Number: 1148-79-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...