Potassium tetrakis(4-chlorophenyl)borate

Supplier: ADIPOGEN CORP MS

Synonyms: Potassiumtetrakis(4-chlorophenyl)borate

CDX-P0134-G001 CDX-P0134-G005
102990-868EA 148.15 USD
102990-868 102990-870
Potassium tetrakis(4-chlorophenyl)borate
Potassium tetrakis(4-chlorophenyl)borate

Lipophilic anionic additive, used together with selective ionophores. The ionophore is the ion carrier which reversibly binds ions and transports them across the hydrophobic membrane. The ionic additive K-TCPB, catalyzes the ion-exchange process at the sample/membrane interface.

Formula: C₂₄H₁₆BCl₄K
MDL Number: MFCD00043105
CAS Number: 14680-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR