7-Methoxycoumarin-4-acetic acid

Supplier: ADIPOGEN CORP MS

CDX-M0028-M500 CDX-M0028-G001
102990-402EA 157.85 USD
102990-402 102990-404
7-Methoxycoumarin-4-acetic acid

Florescent probe that is commonly used as a building block to synthesize coumarin-type fluorescent probes. Spectral data: lambdaex 320 nm, lambdaem 380 nm in methanol. It is used as a fluorescent label for peptides. It is widely used for developing FRET peptide substrates for analyzing protease activities and for HPLC derivatization by fluorescence detection.

Formula: C₁₂H₁₀O₅
MDL Number: MFCD00009774
CAS Number: 62935-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR