Fluorescein Isothiocyanate (mixture of 5- and 6- isomers)

Supplier: ADIPOGEN CORP MS

CDX-F0010-M100 CDX-F0010-G001
102987-938EA 98.31 USD
102987-938 102987-940
Fluorescein Isothiocyanate (mixture of 5- and 6- isomers)
Fluorescein isothiocyanate (mixture of 5 and 6 isomers)

Fluorescein isothiocyanate (FITC) remains one of the most popular fluorescent labeling reagents. Oligonucleotide conjugates of FITC are frequently employed as hybridization probes. Peptide conjugates of FITC and other fluorescent isothiocyanates are susceptible to Edman degradation, making them useful for high-sensitivity amino acid sequencing, FITC-labeled amino acidd and peptides have been separated by capillary electrophoresis with a detection limit of fewer than 1000 molecules. FITC has also been used to detect proteins in gels and on nitrocellulose membranes and is a selective inhibitor of several membrane ATPases.

Formula: C₂₁H₁₁NO₅S
MDL Number: MFCD00005063
CAS Number: 27072-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR