trans-1,3-Diphenyl-2-propen-1-ol

Supplier: ADIPOGEN CORP MS

Synonyms: (E)-1,3-Diphenylprop-2-en-1-ol

CDX-D0106-G025 CDX-D0106-G100
102987-812EA 106.62 USD
102987-812 102987-814
trans-1,3-Diphenyl-2-propen-1-ol
trans-1,3-Diphenyl-2-propen-1-ol

Building block for synthesis.

Formula: C₁₅H₁₄O
MDL Number: MFCD00239426
CAS Number: 62668-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR