Diphenyliodonium hexafluorophosphate

Supplier: ADIPOGEN CORP MS

CDX-D0045-G005 CDX-D0045-G025 CDX-D0045-G100
102987-712EA 105.23 USD
102987-712 102987-714 102987-716
Diphenyliodonium hexafluorophosphate
Diphenyliodonium hexafluorophosphate

Catalyst for the photochemical polymerization of various monomers.

Formula: C₁₂H₁₀F₆IP
MDL Number: MFCD00061398
CAS Number: 58109-40-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR