7-(Diethylamino)coumarin-3-carboxylic acid, N-succinimidyl ester

Supplier: ADIPOGEN CORP MS

Synonyms: 2,5-Dioxopyrrolidin-1-yl 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylate, DEAC, DNP-X acid, SE

CDX-D0037-M100 CDX-D0037-G001
102987-694EA 274.15 USD
102987-694 102987-696
7-(Diethylamino)coumarin-3-carboxylic acid, N-succinimidyl ester
7-(Diethylamino)coumarin-3-carboxylic acid, N-succinimidyl ester

Derivatives of 7-aminocoumarins are the most extensively utilized labeling reagents for preparing brightly blue fluorescent conjugates of proteins and nucleic acid. Labeled amino acids are detected with visible semiconductor laser fluorometry using second harmonic emission (415 nm) of the near-infrared semiconductor laser. Arginine, proline, serine and glycine have been detected with detection limits around 100 amol levels. The fluorescent-labelling better matches the excitation-wavelength of standard blue lasers than conventional fluorescein-isothiocyyanate.

Formula: C₁₈H₁₈N₂O₆
MW: 358.35 g/mol
MDL Number: MFCD00467585
CAS Number: 139346-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR