Cinnamyltriphenylphosphonium chloride

Supplier: ADIPOGEN CORP MS

CDX-C0156-G050 CDX-C0156-G100
102989-508EA 299.08 USD
102989-508 102989-510
Cinnamyltriphenylphosphonium chloride
Cinnamyltriphenylphosphonium chloride

Building block for synthesis. Used in Wittig reactions.

Formula: C₂₇H₂₄ClP
CAS Number: 1530-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR