7-Amino-4-methylcoumarin

Supplier: ADIPOGEN CORP MS

Synonyms: 7-Amino-4-methyl-2-benzopyrone, Coumarin 440, Coumarin 120, AMC

CDX-A0021-M250 CDX-A0021-G010
102989-132EA 113.54 USD
102989-132 102989-134
7-Amino-4-methylcoumarin
7-Amino-4-methylcoumarin

Widely used fluorophore well-known for preparing fluorogenic 7-Amido-4-methylcoumarin (AMC) substrates for cystine aminopeptidase and other hydrolases. Used as reference compound in enzyme assays.

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 352
Melting Pt: 226
MDL Number: MFCD00006868
CAS Number: 26093-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR