DETA NONOate (NOC-18)
Supplier: ADIPOGEN CORP MS
Synonyms:
DETA/NO, 2,2′-(Hydroxynitrosohydrazino)bis-ethanamine, Diethylenetriamine NONOate, 3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene
AG-CR1-3614-M005
AG-CR1-3614-M025
102988-770EA
41.54
USD
102988-770
102988-772
DETA NONOate (NOC-18)
DETA NONOate (NOC-18)
Nitric oxide (NO) donor. The slow release of NO allows the use in experiments investigating the effects of prolonged constant NO delivery to cells and tissues. The half-life time is 20 hours at 37°C and 56 hours at 22-25°C in 0.1M phosphate buffer, pH 7.4. Liberates 2 mol of NO per mol of parent compound. Decomposition of NONOates is nearly instantaneous at pH 5.0.
Formula:
C₄H₁₃N₅O₂ |
MDL Number:
MFCD00278808 CAS Number: 146724-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...