Finasteride

Supplier: ADIPOGEN CORP MS

Synonyms: N-tert-Butyl-3-oxo-4-aza-5alpha-androst-1-en-17beta-carboxamide

AG-CR1-3589-M025 AG-CR1-3589-M100 AG-CR1-3589-M500
102988-674EA 27.69 USD
102988-674 102988-676 102988-678
Finasteride
Finasteride

Potent, specific and competitive inhibitor of type II 5alpha-reductase (enzyme which converts testosterone to the more potent 5alpha-dihydrotestosterone). Anticancer compound. Apoptosis modulator. Enhances the action of GABA at GABA(A) receptors, which leads to neurological implications. Inhibit testosterone-induced type I procollagen and TGF-beta1 expression in human scalp dermal fibroblasts in a model of androgenic alopecia. Drug for the treatment of male androgenetic alopecia. Potential role in neuropsychiatric disorders.

Formula: C₂₃H₃₆N₂O₂
MW: 372.55 g/mol
MDL Number: MFCD00869737
CAS Number: 98319-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR