Artesunate
Supplier: ADIPOGEN CORP MS
Synonyms:
4-Oxo-4-(((3R,5aS,6R,8aS,9R,10R,12R,12aR)-3,6,9-trimethyldecahydro-3H-3,12-epoxy[1,2]dioxepino[4,3-i]isochromen-10-yl)oxy)butanoic acid
AG-CN2-0469-M100
AG-CN2-0469-M500
AG-CN2-0469-G001
102988-386EA
69.23
USD
102988-386
102988-388
102988-390
Artesunate
Artesunate
Semi-synthetic derivative of artemisinin. Anti-malaria. Anti-schistosomiasis. Inhibitor of human cytomegalovirus (CMV). Apoptosis inducer. Anti-cancer. Immunosuppressant. Inhibits production of tumor necrosis factor-alpha (TNF-alpha), interleukin-6 (IL-6), monocyte chemotactic protein-1 (MCP-1) and nitric oxide (NO).
Formula:
C₁₉H₂₈O₈ MW: 384.43 g/mol |
MDL Number:
MFCD00866204 CAS Number: 88495-63-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...